Simport™ Polypropylene Non-sterile Microtubes with Lip Seal
Overview
Brand: Simport™
Manufacturer Part Number: T336-2
UNSPSC: 41121702
Code: 46
Additional Details:
Additional Details: Weight: 1.00000kg
Product Code. 11546879
Quantity Price
1 £ 146.0 / Pack of 1000
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description and Specification
Specification
Packaging Quantity 1000Pack
Closure Color Polyethylene
Product Type Microtube
Temperature Range (Metric) -196 to +110°C