Learn More
Simport™ Polypropylene Non-sterile Microtubes with Lip Seal
Brand:  Simport™ T336-4
UNSPSC : 41121702
Code : 46
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 11576879
  • £147.00 / Pack of 1000
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications
Specifications
Polyethylene
1000/Pk.
-196 to +110°C