missing translation for 'onlineSavingsMsg'
Learn More

Millipore™ Polypropylene Net Filter

Brand:  Millipore™ PP2504700

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10297910

  • £106.50 / Pack of 100


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

White
25μm
Net Filter
100Pack
360μm
Polypropylene