missing translation for 'onlineSavingsMsg'
Learn More

Millipore™ Polypropylene Net Filter

Brand:  Millipore™ PP8002500

Code : NEW

Additional Details : Weight : 0.10000kg

Product Code. 10136680

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

White
80μm
Net Filter
100Pack
450μm
Polypropylene
Videos