missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Tubes™ 3810X

The easy-to-open, securely closing, Eppendorf Tubes™ 3810X offers comfort and dependability for sample preparation, centrifugation, and storage. One vessel for all applications in molecular biology, chemistry, and biochemistry. Safer than ever: designed for centrifugation up to 30,000 x g.

Brand:  Eppendorf™ 0030125207

Code : 96

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 10596901

  • £65.75 / Pack of 1000
In Stock
to see stock.
Add to basket

Description

Description

Use these tubes for all applications in molecular biology, clinical chemistry and biochemistry. Eppendorf™ 3810X Polypropylene Microcentrifuge Tube offers comfort and dependability for sample preparation, centrifugation and storage.

  • Tube lid is easy to open and close for ergonomic operation.
  • Polypropylene provides high resistance to chemicals, mechanical stress, and temperature extremes.
  • g-Safe™: Exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples.
  • Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.
  • Graduation for visual control of the pipetted volume.
  • Frosted lid and surface on the side for easy labeling.
  • Ensured functionality from -86°C to 100°C.
  • Autoclavable when open (121°C, 20 min).
  • Available in Eppendorf Quality and lot-certified PCR clean purity grade.
Specifications

Specifications

Microcentrifuge Tube
Autoclavable
Yellow
1000 Pk.
-86°C to +100°C
Eppendorf Quality
0.05 oz.
Cylindrical
Conical
30,000 x g
1.5 mL
Polypropylene
-122.8°F to +212°F
Non-sterile
Yes
Flat Frost Lid
Certified Quality
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers