Learn More
Millipore™ Polypropylene Microfil Funnel & Filter
Brand: Millipore™ MIRAWG250
Code : 72
Additional Details : Weight : 4.50000kg Transport : UN number : 3270 Chem class : 1 Pack group : II
Description
- Convenient, easy to use system for well accepted membrane filtration method conforms to international standards: CEN, Standard Methods, WHO
- Disposable, stackable, light, recyclable plastic Microfil™ funnels eliminate autoclaving and minimise waste
- 'Push fit' Microfil™ funnels tightly seal to manifold supports without clamps or O-rings assuring leak-free operation and uniform micro-organism recoveries
- Novel device lifts membrane after filtration, for easy removal with forceps
- Unique Microfil™ support filters faster than all-glass filter apparatus
- 100mL or 250mL funnels available with convenient accessory dispensers
- Choose conventional sterile membranes or novel EZ-Pak™ membrane cartridges. Select manifold-mounted filtration supports for laboratory use or stand-alone on-site filtration support.
Specifications
White | |
Polypropylene | |
Yeast or mold in difficult-to-filter sample, Clinical Lab, Food and Beverage, Industrial QC , environmental Analysis, Laboratory Filtration | |
150/Pk. |
47mm | |
1.2μm | |
Microfil Funnel & Filter |