missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Polypropylene Membrane Screw Caps

Catalog Number p-8011598
This item is not returnable. View return policy
This item is not returnable. View return policy
Closure Size GL 25
Diameter (Metric) 33 mm
For Use With (Equipment) Pressure Equalization
Height (Metric) 19 mm
Product Type Membrane Screw Cap
Color Blue
Material Polypropylene
Quantity 5Pack
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'