missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Polypropylene Long Stem Funnels

Brand:  Kartell™ 164

Code : 02

Additional Details : Weight : 0.01000kg

Product Code. 12304026

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

8mm
Suitable for foodstuff
100mL
PP (Polypropylene)
78.5mm
5/Pk.
206mm
Videos