missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Polypropylene Long Stem Funnels

Brand:  Kartell™ 165

Code : 02

Additional Details : Weight : 1

Product Code. 12314026

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

8mm
Suitable for foodstuff
194mm
103.5mm
225mL
PP (Polypropylene)
Videos