missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Polypropylene 'L' Shaped Untapered Connectors

Brand:  Kartell™ 71250

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11914359

  • £7.20 / Pack of 10


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

CE IVD
Natural
Yes
L Untapered Connector
1.3mm Bore
L
4mm
Polypropylene
10/Pk.
Videos