missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Polypropylene 'L' Shaped Untapered Connectors

Brand:  Kartell™ 71256

Code : A0

Additional Details : Weight : 0.04000kg

 View more versions of this product

Product Code. 12579766

  • £9.40 / Pack of 10


In Stock 9
Add to basket

Specifications

Specifications

CE IVD
16mm
Polypropylene
10/Pk.
L
Natural
Yes
L Untapered Connector
11.6mm Bore
Videos