Learn More
Kartell™ Polypropylene 'L' Shaped Untapered Connectors
The ideal leak-proof solution for connecting tubes. Kartell™ Polypropylene 'L' Shaped Untapered Connectors are useful for connecting tubes of various diameters.
Brand: Kartell™ 0125500
Code : A0
Additional Details : Weight : 0.01000kg
Description
Autoclavable. Meet Food and Agriculture regulations. Dimensions are nominal.Specifications
CE IVD | |
14 mm | |
Polypropylene | |
100 pcs. | |
10.5mm Bore |
Natural | |
Autoclavable | |
L Untapered Connector | |
10.5mm Bore | |
L |