missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Polypropylene Graduated Wide Mouth Bottles

Manufactured from polypropylene for excellent chemical and thermal stability. Kartell™ Polypropylene Graduated Wide Mouth Bottles has a wide mouth for easy filling and emptying of liquid or powder samples.

Brand:  Kartell™ 1623

Code : 02

Additional Details : Weight : 0.28576kg

Product Code. 12801304

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Autoclavable
  • Sturdy, rupture-proof bottle
  • Long neck thread ensures leakproof closure
  • Conforms to DIN 13316 and 168
  • Suitable for foodstuff 
Specifications

Specifications

100mL
24mm
1 Each
Yes
20mL
Wide
DIN 13316 and 168
Polypropylene
Yes
GL32
105mm
Videos