missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Plastilab™ Polypropylene Graduated Narrow Neck Bottles

For transporting toxic waste waters, boiler feed water, etc, for analysis as samples.

Brand:  Kartell™ 1603

Code : NEW

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10328294

  • £3.20 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Autoclavable
  • Heavy-walled bottle
  • Secured with shoulder and cap tag
  • Suitable for food stuff
Specifications

Specifications

1000mL
Yes
Polypropylene
Bottle, Narrow Neck
100mL
DIN 13316 and 168
Transporting toxic waste waters, boiler feed water
23mm
Bottle
Narrow
Videos