missing translation for 'onlineSavingsMsg'
Learn More

Polypropylene glycol 4,000, Thermo Scientific Chemicals

Catalog Number p-7051228
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
25322-69-4
(C3H6O)n
134.18
MFCD00084446
DNIAPMSPPWPWGF-UHFFFAOYNA-N
polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol
32881
2-(2-hydroxypropoxy)propan-1-ol
[H]OC(C)CO
This item is not returnable. View return policy
25322-69-4
(C3H6O)n
134.18
MFCD00084446
DNIAPMSPPWPWGF-UHFFFAOYNA-N
polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol
32881
2-(2-hydroxypropoxy)propan-1-ol
[H]OC(C)CO

Propylene glycol is used in the plastics industry for the manufacture of polyester fibers and alkyd resins. It is used as a main ingredient in automobile antifreeze and engine-cooling liquids and in brake and hydraulic fluids. It is used as a humectant in foods and cosmetics. It is used as a solvent for coloring or flavoring agents as well as in many oral, injectable or topical pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Solubility Information Soluble in ethyl alcohol,ethyl ether.
Quantity 50 g
Chemical Name or Material Polypropylene glycol 4,000

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'