Learn More
Kartell™ Polypropylene Filter Tip
Incase of over-aspiration these tips feature orifices that are sized to completely empty the tip. Kartell™ Micropipette Filter Tip eliminates cross-contamination of samples and is compatable with most micropipette brands.
Brand:  Kartell™ 0963900
Code : NEW
Additional Details : Weight : 0.10000kg
Product Code. 10796722
  • £194.00 / Pack of 960
Estimated Shipment: 22-05-2024
to see stock.
Description
Description
  • Includes a filter
  • DNA-free
  • RNA-free
  • Cadmium-free
  • Apyrogenic
  • Sterilized by gamma radiation
  • Autoclavable at 121°C for 20 minutes
  • Designed for use in molecular biology, cell cultures, bacteriology, virology, immunology and radioimmunology