missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Double Open End Carbon-Impregnated Pleated Filter Cartridge, polypropylene
Brand: Cole-Parmer™ 29830-00
Code : A0
Additional Details : Weight : 0.80000kg
Specifications
2 to 3 GPM | |
0.2 μm | |
Filter Cartridge |
Polypropylene | |
1 Pack | |
254 mm |