missing translation for 'onlineSavingsMsg'
Learn More

3M™ Polypropylene Filter Bag

Brand:  3M™ 128A

Code : 04

Additional Details : Weight : 1.70000kg

 View more versions of this product

Product Code. 12368199

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

179mm
Amine Sweetening,‎ Completion & Work over Fluids,‎ Electrodeposition Paint,‎ Electrodeposition Paint Filtration,‎ High Quality Paint Filtration
Polypropylene
813mm
813mm
Filter Bag
Videos