Learn More
CPC Colder Products Company™ HFC Series Polypropylene Coupling Connectors
Constructed from polypropylene. CPC Colder Products Company™ HFC Series Polypropylene Coupling Connectors replaces bulky and heavy ball-and-sleeve couplings in a wide range of applications. The efficient design leads to high flow and low spillage.
Brand: CPC Colder Products Company™ 16612
Code : A0
Additional Details : Weight : 0.80000kg
Description
- High-efficiency valve
- Ergonomic design
- Polypropylene material
- Compatible
- Mates with HFC35 and HFC57