missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Graduated Conical Measures

Manufactured from polypropylene for durability and good chemical resistance. Kartell™ Graduated Conical Measures provide a reliable solution for measuring and dispensing liquids.

Brand:  Kartell™ 423

Code : NEW

Additional Details : Weight : 0.04512kg

 View more versions of this product

Product Code. 12087012

  • £4.35 / Each


In Stock 3
Add to basket

Description

Description

  • Conical shape with integral notch facilitates accurate measurements and easy pours
  • Wide base creates stability
  • Permanent molded graduations provide clarity and reproducibility
  • Polypropylene construction withstands temperatures up to 120°C 
Specifications

Specifications

Yes
Suitable for foodstuff
118mm
120°C
PP (Polypropylene)
Transparent
Yes
2mL
Beaker
Videos