Brand™ Polypropylene Conical Ground Joint Stopper
Overview
Brand: Brand™
Manufacturer Part Number: 144415
UNSPSC: 41122502
Code: 03
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 10218071
Quantity Price
1 £ 1.55 / Each
Estimated Shipment
24-05-2017
Add to basket
Description and Specification
Specification
Product Type Conical Ground Joint Stopper
Size NS12/21
Autoclavable Yes
Bottom Shape Conical
Quantity 1Each
Type Conical Ground Joint Stopper
Material Polypropylene
Shape Square