missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Polypropylene Adapter Housing
Shop All Brand Products
Click to view available options
For Use With (Equipment):
accujet™ pro
Unit Size:
Each
Description
- Polypropylene construction
- For use with accujet™ pro
Specifications
Specifications
| Color | Royal Blue |
| Material | Polypropylene |
| For Use With (Equipment) | accujet™ pro |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction