missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ KleenGuard™ A71 Chemical Spray Protection Coveralls

Catalog Number 12651396
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Kimberly-Clark™ KleenGuard™ A71 Chemical Spray Protection Coveralls create a spray-tight and liquid-tight barrier against harmful substances when working with hazardous chemicals.

  • Approved as Complex Design (Category 3) equipment - offering protection to the levels specified for Type 3, 4 and 5 by CEN
  • Highly visible yellow colour - for improved worker safety
  • Ideal for working with hazardous chemicals - protecting against liquid, aerosol and dry chemical splashes, sprays and dusts in low-pressure industrial cleaning and maintenance areas
  • Single-use protective clothing to improve safety and comfort in high-risk areas - create a spray and liquid tight barrier against harmful chemical splashes, liquids or dust particles
  • Suitable for use as protection against Infective Agents and Biohazards
  • Suitable for use as protection against solid particles and particulate radioactive contamination
  • Treated to be antistatic
  • EN 1073-2 approved for radioactive dust protection
  • Anti-static EN 1149-1 for critical areas
TRUSTED_SUSTAINABILITY
Product Type A71 Coveralls
Size 2X-Large
Color Yellow
Cuff Style Elastic
Hood Type Elastic
For Use With (Application) Hazardous Chemicals
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'