missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Polypropelene Screw Caps with Lip Seal
Click to view available options
Color:
Green
Yellow
Specifications
Closure Type | Screw Cap |
For Use With (Equipment) | For reagent and media bottles |
Color | Green |
Description | Screw cap with lip seal |
Material | Polypropylene |
Length (Metric) | 92.5mm |
Quantity | 10/PK |
Thread Size (Metric) | 45mm |
Certifications/Compliance | ISO R1115 and DIN 168 |
Autoclavable | Autoclavable |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'