missing translation for 'onlineSavingsMsg'
Learn More

Polyphenylmethylsiloxane, MW 2500-2700, Thermo Scientific Chemicals

Catalog Number p-7059377
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
9005-12-3
136.23
MFCD00146731
This item is not returnable. View return policy
9005-12-3
136.23
MFCD00146731

A biochemical for proteomics research. A novel coating for solid-phase microextraction. A polyphenylmethylsiloxane for use in skincare, suncare, color cosmetic, hair care, and antiperspirant / deodorant applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Molecular Formula C7H8OSi
Synonym methylphenylsilanone, polyphenylmethylsiloxane, methylphenyl oxo silane, silane, methyloxophenyl, methyl phenyl silanone
Solubility Information Insoluble in water.
Quantity 50 g
InChI Key IXKNJYSAWNJQQY-UHFFFAOYSA-N
SMILES C[Si](=O)C1=CC=CC=C1
IUPAC Name methyl-oxo-phenylsilane
PubChem CID 3032548
Flash Point 300°C (572°F)
Chemical Name or Material Polyphenylmethylsiloxane, M.W. 2,500 to 2,700
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'