missing translation for 'onlineSavingsMsg'
Learn More

Polyoxyethylene bis(amine), M.W. 8,000, Thermo Scientific Chemicals

Catalog Number p-7080245
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
24991-53-5
H2NCH2CH2(OCH2CH2)nNH2
MFCD00132004
PEG-diamine
This item is not returnable. View return policy
24991-53-5
H2NCH2CH2(OCH2CH2)nNH2
MFCD00132004
PEG-diamine

It is used as PEG modifier, its main products include high-qualified intermediates, various of amino acid and APIs. This Polymer used in the preparation of enzyme conjugates soluble in organic solvents. Promising drug carrier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Granular Solid
Solubility Information It is soluble in organic solvents. It is insoluble in water.
Quantity 250 mg
Sensitivity Air Sensitive
Chemical Name or Material Polyoxyethylene bis (amine), M.W. 8,000

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'