missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polymethylhydrosiloxane, Thermo Scientific Chemicals
21.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Specifications
Color | Colorless |
Physical Form | Viscous Liquid |
Linear Formula | (CH3)3SiO[(CH3)HSiO]nSi(CH3)3 |
Density | 1.0000g/mL |
Solubility Information | Solubility in water: practically insoluble. Other solubilities: may react, soluble in most common organic solvents, (should not be used in halocarbon solvent because, of the possibility of hydride-halide interchange, on catalysis or with heating) |
Quantity | 100 g |
Flash Point | 121°C |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Refractive Index | 1.3970 to 1.4010 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'