Learn More
Thermo Scientific™ Polymer Tubing Cutter
Produce flat, 90°, burr-free ends of polymer tubing for HPLC applications using Thermo Scientific™ Polymer Tubing Cutter.
Brand: Thermo Scientific™ A-328
Code : 16
Additional Details : Weight : 0.90720kg
Description
Produce flat, 90°, burr-free ends of polymer tubing for HPLC applications using the Thermo Scientific™ Polymer Tubing Cutter. It is compatible with rigid polymeric tubing and comes with guide holes for 1/16" and 1/8" tubing.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Replacement Blade | |
5 Pk. | |
Polymeric Tubing Cutter |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.