Learn More
Heidolph™ Polymax 1040 (10° Tilt Angle)
Product Code. 17363030 Shop All Heidolph Instruments Products
Change view
Click to view available options
Plug Type:
UK Plug
Unit Size:
Each
Product Code. Plug Type unitSize
17363030 UK Plug Each
1 options
Product Code. 17363030 Supplier Heidolph™ Supplier No. 5434221006
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The Polymax 1040 can be integrated into the incubation system, making temperature controlled applications possible.
  • The 1000 series device can be integrated into the incubation system. This makes temperature-controlled applications with the Polymax 1040 possible.
  • With a load weight of up to 5 kg, all the required samples can be moved on this platform wave shaker. The speed can be adjusted between 2 to 50 rpm and the device can be operated either continuously or with a timer (max. 120 minutes).
TRUSTED_SUSTAINABILITY
Specifications
Voltage 230 V
Plug Type UK Plug