missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Polygoprep™ 1000 to 30 C4

Brand:  Macherey-Nagel™ 711026.1000

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10777842

  • £2605.00 / 1000g
Estimated Shipment: 16-05-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

25 to 40 μm
POLYGOPREP 1000 to 30
1 kg
Polygoprep
1000 Å
2 to 8
POLYGOPREP C4
Liquid Chromatography
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers