missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Polygoprep™ 1000 to 30 C4
Brand: Macherey-Nagel™ 711026.100
Additional Details : Weight : 0.01000kg
Specifications
25 to 40 μm | |
POLYGOPREP 1000 to 30 | |
0.1 kg | |
Polygoprep |
1000 Å | |
2 to 8 | |
POLYGOPREP C4 | |
Liquid Chromatography |