missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Polygoprep™ 1000 to 30 C4

Brand:  Macherey-Nagel™ 711027.1000

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10072821

  • £3272.85 / 1000g

Estimated Shipment
11-12-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

40 to 63 µm
POLYGOPREP 1000 to 30
1 kg
Polygoprep
1000 Å
2 to 8
POLYGOPREP C4
Liquid Chromatography
Videos
Documents