missing translation for 'onlineSavingsMsg'
Learn More

Polygodial, Tocris Bioscience™

TRPA1 channel activator; analgesic and antifungal

Brand:  Tocris (Bio-Techne) 319710

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13452404

  • £179.34 / 10mg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

TRPA1 channel activator (EC50 = 400 nM) that is selective against TRPV1, TRPV2, TRPV3, TRPV4 and TRPM8 channels. Acts as an analgesic via desensitization of sensory neurons. Also exhibits antifungal activity via inhibition of mitochondrial ATPase.
Specifications

Chemical Identifiers

6754-20-7
234.339
polygodial, tadeonal, poligodial, --polygodial, tadeodal, drim-7-ene-11,12-dial, --tadeonal, unii-5faf7t66m7, 1,4,4a,5,6,7,8,8a-octahydro-5,5,8a-trimethyl-1,2-naphthalenedicarboxaldehyde, 1r,4as,8as-5,5,8a-trimethyl-1,4,4a,5,6,7,8,8a-octahydronaphthalene-1,2-dicarbaldehyde
CHEBI:8305
CC1(CCCC2(C1CC=C(C2C=O)C=O)C)C
C15H22O2
AZJUJOFIHHNCSV-KCQAQPDRSA-N
72503
(1R,4aS,8aS)-5,5,8a-trimethyl-1,4,4a,6,7,8-hexahydronaphthalene-1,2-dicarbaldehyde

Specifications

10mg
6754-20-7
polygodial, tadeonal, poligodial, --polygodial, tadeodal, drim-7-ene-11,12-dial, --tadeonal, unii-5faf7t66m7, 1,4,4a,5,6,7,8,8a-octahydro-5,5,8a-trimethyl-1,2-naphthalenedicarboxaldehyde, 1r,4as,8as-5,5,8a-trimethyl-1,4,4a,5,6,7,8,8a-octahydronaphthalene-1,2-dicarbaldehyde
CC1(CCCC2(C1CC=C(C2C=O)C=O)C)C
234.339
CHEBI:8305
>97%
Polygodial
C15H22O2
AZJUJOFIHHNCSV-KCQAQPDRSA-N
(1R,4aS,8aS)-5,5,8a-trimethyl-1,4,4a,6,7,8-hexahydronaphthalene-1,2-dicarbaldehyde
72503
234.33
Videos
Documents