Learn More
Alfa Aesar™ Polyethyleneimine on silica beads, anion exchange resin, 40-200 mesh
For the separation of nucleotide mixtures
Brand:  Alfa Aesar™ 043303.AB
Code : NEW
Additional Details : CAS Number : 9002-98-6 Weight : 0.01000kg
Quantity 10mL
 View more versions of this product
Product Code. 11379448
  • £17.91 / 10mL
Estimated Shipment date
from Supplier 30-05-2018
Add to basket
Specifications
Chemical Identifiers
Polyethyleneimine on silica beads anion exchange resin
9002-98-6
MFCD00084427
Specifications
Polyethyleneimine on silica beads anion exchange resin
Powder
40 to 200 Mesh
10mL
9002-98-6
MFCD00084427
Hygroscopic