Learn More
Alfa Aesar™ Polyethyleneimine on silica beads, anion exchange resin, 40-200 mesh
For the separation of nucleotide mixtures
Brand:  Alfa Aesar™ 043303.AK
UNSPSC : 12352001
Code : NEW
Additional Details : CAS Number : 9002-98-6 Weight : 0.01000kg
Quantity 250mL
 View more versions of this product
Product Code. 11369448
  • £166.53 / 250mL
Estimated Shipment date
from Supplier 10-07-2017
Add to basket
Specifications Description & Specifications
Chemical Identifiers
Polyethyleneimine on silica beads anion exchange resin
9002-98-6
MFCD00084427
Specifications
Polyethyleneimine on silica beads anion exchange resin
Powder
40 to 200 Mesh
250mL
9002-98-6
MFCD00084427
Hygroscopic
Documentation
Technical Documents