missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Scienceware™ Polyethylene Wide Mouth Wash Bottle

Manufactured from polypropylene. Bel Art™ Polyethylene Wide Mouth Wash Bottle feature a wide mouth 53mm (2 in.) for easy refilling.

Brand:  Bel-Art™ Scienceware™ F11716-0013

Code : 41

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12368118

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

500mL
Saline Solution
Translucent
Natural Polypropylene Cap
4Pack
Compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Wide
Compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Polypropylene
53mm
Polyethylene
Bottle, Wash
Natural
Natural polypropylene cap
Videos
Documentation