missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Polyethylene Wide Mouth Two Color Safety Label Wash Bottle

Catalog Number p-8007858
Click to view available options
Chemical Label:
Acetone
Ethyl Acetate
Methanol
Water
This item is not returnable. View return policy
This item is not returnable. View return policy

Wide-Mouth wash bottles for easy refilling.

  • Valve eliminates drips — allows dispensing of wash bottle in an upright or inverted position
  • Wide mouth 53mm (2 in.) bottle for easy refilling
  • Polypropylene closure; color-coded to aid in easy identification
  • Translucent, LDPE solvent-resistant
  • Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Closure Material Polypropylene
Closure Size 53 mm
Closure Type Green Polypropylene Cap
Through the Closure Dispensing Yes
Chemical Label Ethyl Acetate
Product Type Wash Bottle
Capacity (Metric) 500 mL
Capacity (English) 16.9 oz.
Color Translucent
Diameter (Metric) Outer Neck 53mm
Includes Green Polypropylene Cap
Material Polyethylene
Quantity 3 Pack
Closure Color Green
Certifications/Compliance Compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Closure Included Yes
Autoclavable Not Autoclavable
Graduated No
Mouth Wide
Shape Cylindrical
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'