Bel-Art™ Scienceware™ Polyethylene Wide Mouth Four Color Safety Label Wash Bottle with Vented Cap

Allows dispensing of wash bottle in an upright or inverted position. Bel-Art Products™ Polyethylene Wide Mouth Four Color Safety Label Wash Bottle with Vented Cap's color-coded polypropylene closures aid in quick and easy identification.

Overview
Brand: Bel-Art™ Scienceware™

Manufacturer Part Number: F11808-0004

UNSPSC: 41103209

Code: 41

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12348128

Quantity Price
1 £ 32.34 / Pack of 4
Estimated Shipment
24-05-2017
Add to basket

Wash bottles in this range that are prelabelled for intended usage with specific hazardous substances are also prelabelled with the appropriate GHS hazard warning symbols.
 

  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification

Specification

Closure Color Blue
Type Wash bottle
Closure Type Cap, Safety Vented
Mouth Wide
Certifications/Compliance Compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Certifications Compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Diameter (Metric) Outer Mouth 53mm
Quantity 4Pack
Chemical Label Distilled Water
Material Polyethylene
Color Translucent
Includes Blue Polypropylene Cap
Capacity (Metric) 250mL
Closure Material Polypropylene

  • Patented valve eliminates drips and allows bottle to be used upright or inverted
  • Full color NFPA diamond
  • Colored closures for enhanced identification
  • LDPE bottles with polypropylene caps
  • Sodium hypochlorite bottles are white for light protection
  • Available in 250mL, 500mL or 1L bottles For dispensing water and common solvents.

Alternative Products
Documentation