missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Scienceware™ Polyethylene Wide Mouth Four Color Safety Label Wash Bottle with Vented Cap

Catalog Number 12398128
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Closure Color:
Assorted
Blue
Green
Natural
Red
Yellow
Chemical Label:
Acetone
Custom Write-on
Distilled Water
Ethanol
Ethyl Acetate
Isopropanol
Methanol
Sodium Hypochlorite (Bleach)
This item is not returnable. View return policy
Wash bottles in this range that are prelabelled for intended usage with specific hazardous substances are also prelabelled with the appropriate GHS hazard warning symbols.
 
This item is not returnable. View return policy
Wash bottles in this range that are prelabelled for intended usage with specific hazardous substances are also prelabelled with the appropriate GHS hazard warning symbols.
 

Allows dispensing of wash bottle in an upright or inverted position. Bel-Art™ SP Scienceware™ Polyethylene Wide Mouth Four Color Safety Label Wash Bottle with Vented Cap's color-coded polypropylene closures aid in quick and easy identification.

  • Patented valve eliminates drips and allows bottle to be used upright or inverted
  • Full color NFPA diamond
  • Colored closures for enhanced identification
  • LDPE bottles with polypropylene caps
  • Sodium hypochlorite bottles are white for light protection
  • Available in 250mL, 500mL or 1L bottles For dispensing water and common solvents.
Closure Material Polypropylene
Closure Type Yellow Polypropylene Cap
Chemical Label Sodium Hypochlorite (Bleach)
Product Type Wash Bottle
Capacity (Metric) 250 mL
Color Translucent
Diameter (Metric) Outer Neck 53mm
Includes Yellow Polypropylene Cap
Material Polyethylene
Quantity 4Pack
Closure Color Yellow
Certifications/Compliance Compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Mouth Wide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'