missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Scienceware™ Polyethylene Wide Mouth Four Color Safety Label Wash Bottle with Vented Cap

Allows dispensing of wash bottle in an upright or inverted position. Bel-Art™ SP Scienceware™ Polyethylene Wide Mouth Four Color Safety Label Wash Bottle with Vented Cap's color-coded polypropylene closures aid in quick and easy identification.

Brand:  Bel-Art™ Scienceware™ F11816-0009

Code : 41

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12318138

  • £32.54 / Pack of 4


Call for latest availability
+44 (0)1509 555 500
Add to basket

Wash bottles in this range that are prelabelled for intended usage with specific hazardous substances are also prelabelled with the appropriate GHS hazard warning symbols.
 

Description

Description

  • Patented valve eliminates drips and allows bottle to be used upright or inverted
  • Full color NFPA diamond
  • Colored closures for enhanced identification
  • LDPE bottles with polypropylene caps
  • Sodium hypochlorite bottles are white for light protection
  • Available in 250mL, 500mL or 1L bottles For dispensing water and common solvents.
Specifications

Specifications

500mL
LYOB (Label Your Own Bottle)
Translucent
Natural Polypropylene Cap
4Pack
Compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Wide
Compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Polypropylene
53mm
Polyethylene
Bottle, Wash
Natural
Natural polypropylene cap
Videos
Documentation