Bel-Art™ Scienceware™ Polyethylene Wide Mouth Four Color Safety Label Wash Bottle with Vented Cap

Allows dispensing of wash bottle in an upright or inverted position. Bel-Art Products™ Polyethylene Wide Mouth Four Color Safety Label Wash Bottle with Vented Cap's color-coded polypropylene closures aid in quick and easy identification.

Overview
Brand: Bel-Art™ Scienceware™

Manufacturer Part Number: F11832-0019

UNSPSC: 41103209

Code: 41

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12358138

Quantity Price
1 £ 45.63 / Pack of 2
Call for latest availability
+44 (0)1509 555 500
Add to basket

Wash bottles in this range that are prelabelled for intended usage with specific hazardous substances are also prelabelled with the appropriate GHS hazard warning symbols.
 

Description and Specification

Specification

Certifications Compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Diameter (Metric) Outer Mouth 53mm
Closure Natural polypropylene cap
Closure Type Cap, Safety Vented
Quantity 2Pack
Product Type Wash bottle
Chemical Label Ethanol
Material Polyethylene
Color Translucent
Bottle Material Polyethylene
Closure Color Natural
Includes Natural Polypropylene Cap
Capacity (Metric) 1000mL
Closure Material Polypropylene
Wide Mouth Yes
Mouth Style Wide

  • Patented valve eliminates drips and allows bottle to be used upright or inverted
  • Full color NFPA diamond
  • Colored closures for enhanced identification
  • LDPE bottles with polypropylene caps
  • Sodium hypochlorite bottles are white for light protection
  • Available in 250mL, 500mL or 1L bottles For dispensing water and common solvents.

Documentation