Learn More
Bel-Art™ Scienceware™ Polyethylene Wide Mouth Four Color Safety Label Wash Bottle with Vented Cap
Allows dispensing of wash bottle in an upright or inverted position. Bel-Art™ SP Scienceware™ Polyethylene Wide Mouth Four Color Safety Label Wash Bottle with Vented Cap's color-coded polypropylene closures aid in quick and easy identification.
Brand: Bel-Art™ Scienceware™ F11808-0019
Code : 41
Additional Details : Weight : 0.01000kg
Description
- Patented valve eliminates drips and allows bottle to be used upright or inverted
- Full color NFPA diamond
- Colored closures for enhanced identification
- LDPE bottles with polypropylene caps
- Sodium hypochlorite bottles are white for light protection
- Available in 250mL, 500mL or 1L bottles For dispensing water and common solvents.
Specifications
250mL | |
Ethanol | |
Translucent | |
Natural Polypropylene Cap | |
4Pack | |
Compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200 | |
Wide |
Compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200 | |
Polypropylene | |
53mm | |
Polyethylene | |
Bottle, Wash | |
Natural | |
Natural polypropylene cap |