Kartell™ Polyethylene Short Spout Dropping Bottle Screw Caps
Overview
Brand: Kartell™
Manufacturer Part Number: 3304
UNSPSC: 24122004
Code: NEW
Additional Details:
Additional Details: Weight: 0.10000kg
Product Code. 10768302
Quantity Price
1 £ 31.15 / Pack of 50
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description and Specification
Specification
Type Short Spout Dropping Bottle Screw Cap
Closure Type Yes
Includes PE cap for tip closure
For Use With (Equipment) Narrow neck standardized bottles part. no. 15801 - 15821 - 15841 - 15861 - 15881 - 15901 and wide neck standardized bottles part. no. 1608 - 1610
Product Line Cap
Screw Material Polyethylene
Closure Material Polyethylene