Learn More
Thermo Scientific Alfa Aesar Polyethylene sheet, Low Density, 4.8mm (0.19 in.) thick
Brand: Thermo Scientific Alfa Aesar 046908.HB
Polyethylene sheet, Low Density, 4.8mm (0.19 in.) thick
Code : NEW
Additional Details : CAS Number : 9002-88-4 Weight : 0.01000kg