missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene sheet, High Density, 3.18mm (0.125in) thick, Thermo Scientific™
Click to view available options
Quantity:
300 x 300 mm
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
CAS | 9002-88-4 |
Molecular Formula | (C2H4)n |
MDL Number | MFCD00084423 |
Synonym | ethylene, acetene, elayl, olefiant gas, athylen, etileno, liquid ethylene, polyethylene, bicarburretted hydrogen, plastipore |
Quantity | 300 x 300 mm |
InChI Key | VGGSQFUCUMXWEO-UHFFFAOYSA-N |
SMILES | *-CC-* |
IUPAC Name | ethene |
Molecular Weight (g/mol) | 28.05 |
PubChem CID | 6325 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'