missing translation for 'onlineSavingsMsg'
Learn More

Azpack Pin adamantine, nickel plated mild steel, 51mm x 1.2mm

Brand:  Azpack XFL-800-011Y

Pin adamantine, nickel plated mild steel, 51mm x 1.2mm

Code : 02

Additional Details : Weight : 0.14040kg

Product Code. 12682590

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.