missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene rod, High Density, 19mm (0.75 in.) dia. 2.5cm, Thermo Scientific™

Catalog Number p-7052205
Click to view available options
Quantity:
2.5 cm
This item is not returnable. View return policy
9002-88-4
(C2H4)n
28.05
MFCD00084423
VGGSQFUCUMXWEO-UHFFFAOYSA-N
ethylene, acetene, elayl, olefiant gas, athylen, etileno, liquid ethylene, polyethylene, bicarburretted hydrogen, plastipore
6325
CHEBI:18153
ethene
*-CC-*
This item is not returnable. View return policy
9002-88-4
(C2H4)n
28.05
MFCD00084423
VGGSQFUCUMXWEO-UHFFFAOYSA-N
ethylene, acetene, elayl, olefiant gas, athylen, etileno, liquid ethylene, polyethylene, bicarburretted hydrogen, plastipore
6325
CHEBI:18153
ethene
*-CC-*

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Physical Form Rod
Quantity 2.5 cm
Odor Odorless
Chemical Name or Material Polyethylene rod, High Density, 19mm (0.75 in.) dia.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'