missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Polyethylene rod, High Density, 16mm (0.63 in.) dia.

Brand:  Alfa Aesar™ 045202.BV

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 30cm

Product Code. 15496815

  • £6.15 / 30cm


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Specifications

Polyethylene rod
9002-88-4
30cm
High Density, 16mm (0.63 in.) dia.
Rod
Videos
SDS