missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene rod, High Density, 12.7mm (0.5 in.) dia., Thermo Scientific™

Catalog Number 15426835
Click to view available options
Quantity:
30 cm
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 9002-88-4
Physical Form Rod
Molecular Formula (C2H4)n
MDL Number MFCD00084423
Synonym ethylene, acetene, elayl, olefiant gas, athylen, etileno, liquid ethylene, polyethylene, bicarburretted hydrogen, plastipore
Quantity 30 cm
InChI Key VGGSQFUCUMXWEO-UHFFFAOYSA-N
SMILES *-CC-*
IUPAC Name ethene
Molecular Weight (g/mol) 28.05
PubChem CID 6325
ChEBI CHEBI:18153
Odor Odorless
Chemical Name or Material Polyethylene rod, High Density, 12.7mm (0.5 in.) dia.
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'