missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Dispolab™ Pasteur Disposable Pipettes

Designed with slender tip to guarantee precision transfer of samples.

Brand:  Kartell™ 88337

Code : NEW

Additional Details : Weight : 10.25000kg

 View more versions of this product

Product Code. 10458842

  • £187.00 / Pack of 5000


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Made from polyethylene
  • With bulb and graduations
  • Sterile pipettes individually wrapped
  • Non-sterile pipettes available in multi-packs
Specifications

Specifications

CE–IVD
Yes
152.19mm
Slender
Pasteur Pipet
PE (Polyethylene)
Yes
0.5mL
500/Pack
Bulb shaped
3mL
Sterile
Videos